366919 Sigma-Aldrich

Chloroform

ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer

DOWNLOAD MSDS (PDF)

Synonym: Methylidyne trichloride, Trichloromethane

Properties

Related Categories Amber Glass Bottles, Chloroform, Solvent by Type, Solvent Bottles, Solvent Packaging Options,
grade   ACS spectrophotometric grade
vapor density   4.1 (vs air)
vapor pressure   160 mmHg ( 20 °C)
assay   ≥99.8%
contains   0.5-1.0% ethanol as stabilizer
impurities   Acetone and aldehyde, passes test
  Acid and chloride, passes test
  Free chlorine (Cl), passes test
  H2SO4, passes test(darkened)
  <0.03% water
evapn. residue   <0.001%
color   APHA: ≤10
refractive index   n20/D 1.445(lit.)
bp   60.5-61.5 °C(lit.)
mp   −63 °C(lit.)
density   1.48 g/mL at 25 °C (represents the value for the density of chloroform with ethanol stabilizer)
  1.492 g/mL at 25 °C(lit.)
cation traces   Pb: ≤0.05 ppm
λ   H2O reference
UV absorption   λ: 245 nm Amax: 1.00
  λ: 255 nm Amax: 0.25
  λ: 260 nm Amax: 0.15
  λ: 270 nm Amax: 0.05
  λ: 290-400 nm Amax: 0.01

Description

Packaging

1, 6×1, 2, 4×4 L in glass bottle

Price and Availability

Customers Also Viewed

Chloroform

ACS spectrophotometric grade, ≥99.8%, contains amylenes as stabilizer

Chloroform

ACS reagent, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer

Chloroform

contains ethanol as stabilizer, ACS reagent, ≥99.8%

Chloroform

contains ~0.5% ethanol as stabilizer, for IR spectroscopy, ≥99.5%

Chloroform

for UV-spectroscopy, ≥99.8% (chloroform + ethanol, GC)

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?