EMAIL THIS PAGE TO A FRIEND
36695 Sigma-Aldrich

2-Chlorotoluene

PESTANAL®, analytical standard

  • CAS Number 95-49-8

  • Linear Formula CH3C6H4Cl

  • Molecular Weight 126.58

  •  Beilstein Registry Number 1904175

  •  EC Number 202-424-3

  •  MDL number MFCD00000562

  •  PubChem Substance ID 329755459

  •  eCl@ss 39050205

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, C, Pesticides & Metabolites Standards More...
grade   analytical standard
vapor density   4.38 (vs air)
vapor pressure   10 mmHg ( 43 °C)
InChI Key   IBSQPLPBRSHTTG-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.525(lit.)
bp   157-159 °C(lit.)
mp   −36 °C(lit.)
solubility   H2O: slightly soluble0.047 g/L at 20 °C
density   1.083 g/mL at 25 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 2238 3 / PGIII
WGK Germany 
2
RTECS 
XS9000000
Flash Point(F) 
107.6 °F
Flash Point(C) 
42 °C
Protocols & Articles

Articles

GC Analysis of Volatiles on SPB®-624, Direct Injection

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles on SPB®-624, Direct Injection
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Volatiles on VOCOL® after SPME using 75 μm Carboxen/PDMS Fiber

From our library of Articles, Sigma-Aldrich presents GC Analysis of Volatiles on VOCOL® after SPME using 75 μm Carboxen/PDMS Fiber
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Solid phase microextractions

US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, ELCD Signal

From our library of Articles, Sigma-Aldrich presents US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, ELCD Signal
Keywords: Chromatography, Gas chromatography, Hall electrolytic conductivity detector, Photo-ionization detector, Purge and trap, Sample preparations

US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, PID Signal

From our library of Articles, Sigma-Aldrich presents US EPA Method 502.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap, PID Signal
Keywords: Chromatography, Gas chromatography, Hall electrolytic conductivity detector, Photo-ionization detector, Purification

US EPA Method 524.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 524.2: GC Analysis of Volatiles on VOCOL® (105 m x 0.53 mm I.D.) after Purge & Trap using "K" Trap
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8260: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on SPB®-624 after Purge & Trap using "K" Trap
Keywords: Chromatography, Environmental, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap

From our library of Articles, Sigma-Aldrich presents US EPA Method 8260: GC Analysis of Volatiles on VOCOL® after Purge & Trap using "K" Trap
Keywords: Chromatography, Gas chromatography, Purification, Solid phase microextractions

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?