EMAIL THIS PAGE TO A FRIEND
36703 Sigma-Aldrich

2,6-Dichloroaniline

PESTANAL®, analytical standard

Purchase

Properties

Related Categories Additional Standards, Alpha Index of All Pesticide Standards, Amines, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
InChI Key   JDMFXJULNGEPOI-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
mp   36-38 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3442 6.1 / PGII
WGK Germany 
3
Flash Point(F) 
244.4 °F
Flash Point(C) 
118 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?