EMAIL THIS PAGE TO A FRIEND
36713 Sigma-Aldrich

2,3-Dimethylphenol

PESTANAL®, analytical standard

Synonym: 3-Hydroxyl-o-xylene, vic.-o-Xylenol

  • CAS Number 526-75-0

  • Linear Formula (CH3)2C6H3OH

  • Molecular Weight 122.16

  •  Beilstein Registry Number 1906267

  •  EC Number 208-395-3

  •  MDL number MFCD00002227

  •  PubChem Substance ID 329755472

Purchase

Properties

Related Categories A-F, A-L, Additional Standards, All Alphabetically Sorted, Alpha Index of All Pesticide Standards,
grade   analytical standard
vapor density   4.23 (vs air)
InChI Key   QWBBPBRQALCEIZ-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   217 °C(lit.)
mp   70-73 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2261 6.1 / PGII
WGK Germany 
3
RTECS 
ZE5500000
Flash Point(F) 
203 °F
Flash Point(C) 
95 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?