EMAIL THIS PAGE TO A FRIEND
36716 Sigma-Aldrich

3,4-Dimethylphenol

PESTANAL®, analytical standard

Synonym: 4-Hydroxy-o-xylene

  • CAS Number 95-65-8

  • Linear Formula (CH3)2C6H3OH

  • Molecular Weight 122.16

  •  Beilstein Registry Number 1099267

  •  EC Number 202-439-5

  •  MDL number MFCD00002304

  •  PubChem Substance ID 329755475

Purchase

Properties

Related Categories A-F, A-L, Additional Standards, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted,
grade   analytical standard
InChI Key   YCOXTKKNXUZSKD-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   227 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 2261 6.1 / PGII
WGK Germany 
3
RTECS 
ZE6300000
Flash Point(F) 
230 °F
Flash Point(C) 
110 °C
Protocols & Articles

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?