EMAIL THIS PAGE TO A FRIEND
36723 Sigma-Aldrich

3-Ethylphenol

PESTANAL®, analytical standard

  • CAS Number 620-17-7

  • Linear Formula C2H5C6H4OH

  • Molecular Weight 122.16

  •  Beilstein Registry Number 1857002

  •  EC Number 210-627-3

  •  MDL number MFCD00002311

  •  PubChem Substance ID 24862405

Purchase

Properties

Related Categories A-F, A-L, Additional Standards, Alcoholic & Nonalcoholic Beverages, All Alphabetically Sorted,
grade   analytical standard
InChI Key   HMNKTRSOROOSPP-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.533(lit.)
density   1.001 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3145 8 / PGIII
WGK Germany 
3
Flash Point(F) 
201.2 °F
Flash Point(C) 
94 °C

Documents

Certificate of Analysis

Protocols & Articles

Related Content

Flavor and Fragrance Analysis

It’s all about your workflow. Sample preparation, separation, detection and analysis. Speed and resolution is key in determining purity or verifying composition.
Keywords: Calorimetry, Gas chromatography, Mass spectrometry, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?