EMAIL THIS PAGE TO A FRIEND
36737 Sigma-Aldrich

Diethyl phthalate

PESTANAL®, analytical standard

  • CAS Number 84-66-2

  • Linear Formula C6H4-1,2-(CO2C2H5)2

  • Molecular Weight 222.24

  •  Beilstein Registry Number 1912500

  •  EC Number 201-550-6

  •  MDL number MFCD00009111

  •  PubChem Substance ID 329755485

Purchase

Properties

Related Categories 2000/60/EC, Additional Standards, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
vapor density   7.66 (vs air)
vapor pressure   1 mmHg ( 100 °C)
InChI Key   FLKPEMZONWLCSK-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
autoignition temp.   854 °F
shelf life   limited shelf life, expiry date on the label
expl. lim.   0.75 %, 187 °F
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.502(lit.)
bp   298-299 °C(lit.)
mp   −3 °C(lit.)
density   1.12 g/mL at 25 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Personal Protective Equipment 
RIDADR 
UN 3082 9 / PGIII
WGK Germany 
2
RTECS 
TI1050000
Flash Point(F) 
312.8 °F
Flash Point(C) 
156 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

53008 Diethyl phthalate, PESTANAL®, analytical standard

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?