EMAIL THIS PAGE TO A FRIEND
36738 Sigma-Aldrich

Dimethyl phthalate

PESTANAL®, analytical standard

Synonym: DMP

  • CAS Number 131-11-3

  • Linear Formula C6H4-1,2-(CO2CH3)2

  • Molecular Weight 194.18

  •  Beilstein Registry Number 1911460

  •  EC Number 205-011-6

  •  MDL number MFCD00008425

  •  PubChem Substance ID 329755486

Purchase

Properties

Related Categories A-F, A-L, Additional Pesticide Standards, Additional Standards, All Alphabetically Sorted,
grade   analytical standard
InChI Key   NIQCNGHVCWTJSM-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
autoignition temp.   1033 °F
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.515(lit.)
bp   282 °C(lit.)
mp   2 °C(lit.)
density   1.19 g/mL at 25 °C(lit.)
format   neat

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
UN 3082 9 / PGIII
WGK Germany 
1
Flash Point(F) 
294.8 °F
Flash Point(C) 
146 °C
Protocols & Articles

Articles

GC Analysis of Dibasic Acid Methyl Esters on SP™-2380

From our library of Articles, Sigma-Aldrich presents GC Analysis of Dibasic Acid Methyl Esters on SP™-2380
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification, Size-exclusion chromatography

GC Analysis of Phthalate Esters on Equity®-5

From our library of Articles, Sigma-Aldrich presents GC Analysis of Phthalate Esters on Equity®-5
Keywords: Chromatography, Flame ionization detector, Gas chromatography, Purification

GC Analysis of Semivolatiles in Drinking Water on a 5% Phenyl Column after SPE using ENVI™-18 DSK

From our library of Articles, Sigma-Aldrich presents GC Analysis of Semivolatiles in Drinking Water on a 5% Phenyl Column after SPE using ENVI™-18 DSK
Keywords: Chromatography, Gas chromatography, Mass spectrometry, Purification, Sample preparations, Solid phase extractions

GC Analysis of Semivolatiles in Water on a 5% Phenyl Column after SPME using 7 μm PDMS

From our library of Articles, Sigma-Aldrich presents GC Analysis of Semivolatiles in Water on a 5% Phenyl Column after SPME using 7 μm PDMS
Keywords: Chromatography, Environmental, Gas chromatography, Mass spectrometry, Purification, Size-exclusion chromatography, Solid phase microextractions

HPLC Analysis of Column Test Mixture on Ascentis® Express RP-Amide

From our library of Articles, Sigma-Aldrich presents HPLC Analysis of Column Test Mixture on Ascentis® Express RP-Amide
Keywords: High performance liquid chromatography

SPME-GC-MS Determination of Phthalate Esters in Ramen Noodle Flavor Packets

In June of 2011, food safety authorities in Hong Kong found phthalate ester contamination in a variety of imported food and drink products1. Among these foods were several brands of ramen noodle kits...
Katherine K. Stenerson
Reporter US, Volume 30.1
Keywords: Chromatography, Food Safety, Gas chromatography, Gas chromatography mass spectrometry, Mass selective detector, Mass spectrometry, Sample preparations, Size-exclusion chromatography, Solid phase microextractions

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 525.2: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 625: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on Equity®-5
Keywords: Chromatography, Gas chromatography, Mass selective detector, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB-5ms (20 m x 0.18 mm I.D., 0.36 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.18 μm), Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification, Size-exclusion chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (20 m x 0.18 mm I.D., 0.36 μm), 28 °C/min Oven Ramp, Fast GC Analysis
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms (30 m x 0.25 mm I.D., 0.50 μm)
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms, 350 °C Final Oven Temp.
Keywords: Chromatography, Gas chromatography

US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method 8270: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents US EPA Method OLM04.2 SVOA: GC Analysis of Semivolatiles on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Purification

Related Content

Flavor and Fragrance Analysis

We offer quality product solutions in all three stages of analysis to help achieve fast, accurate results. Below, you can learn more about our workflow solutions for sample preparation, separation, d...
Keywords: Calorimetry, Environmental, Gas chromatography, Mass spectrometry, Pesticides, Sample preparations, Separation

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?