EMAIL THIS PAGE TO A FRIEND
36759 Sigma-Aldrich

2,2′-Bipyridyl

PESTANAL®, analytical standard

Synonym: 2,2′-Bipyridine, 2,2′-Dipyridine, 2,2′-Dipyridyl

  • CAS Number 366-18-7

  • Empirical Formula (Hill Notation) C10H8N2

  • Molecular Weight 156.18

  •  Beilstein Registry Number 113089

  •  EC Number 206-674-4

  •  MDL number MFCD00006212

  •  PubChem Substance ID 329755497

Purchase

Properties

Related Categories A-B, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Pesticides & Metabolites Standards More...
grade   analytical standard
InChI Key   ROFVEXUMMXZLPA-UHFFFAOYSA-N
product line   PESTANAL®
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
bp   273 °C(lit.)
mp   70-73 °C(lit.)
format   neat
Gene Information   human ... EPHX2(2053), FNTA(2339)
mouse ... Ephx2(13850)

Description

Biochem/physiol Actions

Metalloprotease inhibitor, high-affinity chelator of iron; may inhibit Fe2+ containing enzymes at 10−8 M.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability


Laboratory Gloves
Safety & Documentation

Safety Information

Symbol 
GHS06  GHS06
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 2811 6.1 / PGIII
WGK Germany 
3
RTECS 
DW1750000
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?