EMAIL THIS PAGE TO A FRIEND
36765 Sigma-Aldrich

2,6-Diethylaniline

PESTANAL®, analytical standard

Synonym: 2-Amino-1,3-diethylbenzene

  • CAS Number 579-66-8

  • Linear Formula (C2H5)2C6H3NH2

  • Molecular Weight 149.23

  •  Beilstein Registry Number 1423626

  •  EC Number 209-445-7

  •  MDL number MFCD00007753

  •  PubChem Substance ID 329755501

Purchase

Properties

Related Categories Additional Standards, Alpha Index of All Pesticide Standards, Amines, Analytical Standards, Analytical/Chromatography,
grade   analytical standard
vapor pressure   0.02 mmHg ( 20 °C)
InChI Key   FOYHNROGBXVLLX-UHFFFAOYSA-N
product line   PESTANAL®
assay   98-100% (GC)
form   neat
shelf life   limited shelf life, expiry date on the label
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
refractive index   n20/D 1.545(lit.)
bp   243 °C(lit.)
density   0.906 g/mL at 25 °C(lit.)
format   neat

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2
RTECS 
BX3500000
Flash Point(F) 
239 °F
Flash Point(C) 
115 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?