• USA Home
  • 36775 - 2′,3′-Dideoxycytidine

EMAIL THIS PAGE TO A FRIEND
36775 Sigma-Aldrich

2′,3′-Dideoxycytidine

≥99.0% (HPLC)

Synonym: ddC

  • CAS Number 7481-89-2

  • Empirical Formula (Hill Notation) C9H13N3O3

  • Molecular Weight 211.22

  •  Beilstein Registry Number 654956

  •  MDL number MFCD00012188

  •  PubChem Substance ID 329755507

Purchase

Description

Biochem/physiol Actions

Infection of NIH Swiss 3T3 cells by 334C murine leukemia virus is inhibited by 50 μM ddCyd without effecting cell growth; ddCyd inhibits the in vitro infectivity and cytopathic effects of human Tlymphotropic virus type III/ lymphadenopathy-associated virus (HTLV-III/LAV).

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS08  GHS08
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
HA3870000

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?