• USA Home
  • 36809 - 3,3′-Diethylthiacarbocyanine iodide

EMAIL THIS PAGE TO A FRIEND
36809 Sigma-Aldrich

3,3′-Diethylthiacarbocyanine iodide

for fluorescence, ≥98.0% (HPCE)

Synonym: 3-Ethyl-2-(3-[3-ethyl-2(3H)benzothiazolylidene]-1-propenyl)benzothiazolium iodide, DiSC2(3)

  • CAS Number 905-97-5

  • Empirical Formula (Hill Notation) C21H21IN2S2

  • Molecular Weight 492.44

  •  Beilstein Registry Number 4117818

  •  MDL number MFCD00011956

  •  PubChem Substance ID 329755514

Purchase

Properties

Related Categories Alphabetical, Biochemicals and Reagents, Detection, Dialkylcarbocyanine and Styryl probes, Fluorescent Indicators and Probes,
grade   for fluorescence
InChI Key   VZBILKJHDPEENF-UHFFFAOYSA-M
assay   ≥98.0% (HPCE)
mp   268-270 °C (dec.)(lit.)
solubility   DMSO: soluble
  methanol: soluble
fluorescence   λex 554 nm; λem 577 nm in phosphate buffer/SDS pH 7.0

Description

Application

3,3′-Diethylthiacarbocyanine iodide [DiSC2(3)], a light sensitive carbocyanine dye, is used as a photobleaching agent in peptide nucleic acid (PNA)-DNA duplex research.

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
RTECS 
DL7061500

Documents

Certificate of Analysis

Protocols & Articles

Articles

Sensor Dyes

In 1998 R.F. Furchgott, L.J. Ignarro, and F. Murad received the Nobel Prize in Physiology and Medicine for their discoveries concerning nitric oxide as a signaling molecule in the cardiovascular syst...
Monika Baeumle
BioFiles 2011, 6.3, 23.
Keywords: Applications, Biochemistry, Biological processes, Cancer, Cardiovascular, Central Nervous System, Diseases, Extinction coefficient, Indicators, Inflammation, Methods, Microscopy, Neurotransmission, Sol-gel, Type

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?