• USA Home
  • 368458 - Aluminum sulfate hydrate

EMAIL THIS PAGE TO A FRIEND
368458 Sigma-Aldrich

Aluminum sulfate hydrate

98%

Purchase

Properties

Related Categories Aluminum Salts, Essential Chemicals, Research Essentials, Salts
assay   98%
composition   Degree of hydration, 14-18
density   1.69 g/mL at 25 °C(lit.)

Description

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Packaging

2.5 kg in poly bottle

500 g in poly bottle

Application

Aluminum sulfate hydrate was used in the synthesis of new high crystalline zeolite materials.
It may be used in the synthesis of Al-doped ZnO nanorods and spherical alumina powders.

General description

Aluminum sulfate hydrate is a hydrated aluminum salt. Aluminum sulfate is widely used in oil and fat processing, paper manufacturing and as a flocculant for water treatment. The kinetics of its dehydration by thermal decomposition has been investigated. It is one of the components of hematoxylin solution. Spherical particles of aluminum sulfate hydrate has been synthesized by prilling technique.

Price and Availability

Suggested Laboratory Gloves


Laboratory GlovesThis substance has been tested against several types of hand protection for CE compliance. Click below to find the recommended gloves for handling this product.



Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
UN 3077 9 / PGIII
WGK Germany 
1

Documents

Certificate of Analysis

Certificate of Origin

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?