EMAIL THIS PAGE TO A FRIEND
36881 Sigma-Aldrich

3,5-Difluoroaniline

purum, ≥98.0% (GC)

  • CAS Number 372-39-4

  • Linear Formula F2C6H3NH2

  • Molecular Weight 129.11

  •  Beilstein Registry Number 2802286

  •  EC Number 206-752-8

  •  MDL number MFCD00007763

Purchase

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3
Flash Point(F) 
167 °F
Flash Point(C) 
75 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

263532 3,5-Difluoroaniline, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?