EMAIL THIS PAGE TO A FRIEND
36905 Sigma-Aldrich

PCB No 180 solution

10 μg/mL in isooctane, analytical standard

Synonym: 2,2′,3,4,4′,5,5′-Heptachlorobiphenyl solution, 2,2′,3,4,4′,5,5′-PCB

  • CAS Number 35065-29-3

  • Empirical Formula (Hill Notation) C12H3Cl7

  • Molecular Weight 395.32

  •  Ballschmiter Number 180

  •  Beilstein Registry Number 2509255

  •  EC Number 208-759-1

  •  MDL number MFCD00055551

  •  PubChem Substance ID 329755533

Purchase

Properties

Related Categories Additional Standards, Analytical Standards, Analytical/Chromatography, Aroclors, PCBs, and Dioxins, Chromatography,
grade   analytical standard
InChI Key   WBHQEUPUMONIKF-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
concentration   10 μg/mL in isooctane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1262 - class 3 - PG 2 - Octanes
WGK Germany 
2
Flash Point(F) 
10.4 °F
Flash Point(C) 
-12 °C
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?