• USA Home
  • 36935 - Octachloronaphthalene solution

EMAIL THIS PAGE TO A FRIEND
36935 Sigma-Aldrich

Octachloronaphthalene solution

10 μg/mL in cyclohexane, PESTANAL®, analytical standard

  •  Beilstein Registry Number 1653604

  •  MDL number MFCD00145026

Purchase

Properties

Related Categories Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Aryl, Building Blocks,
grade   analytical standard
InChI Key   RTNLUFLDZOAXIC-UHFFFAOYSA-N
product line   PESTANAL®
concentration   10 μg/mL in cyclohexane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Other Notes

Same product — same quality — now Sigma brand

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1145 - class 3 - PG 2 - Cyclohexane, solution
WGK Germany 
2
Flash Point(F) 
-.4 °F
Flash Point(C) 
-18 °C

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?