EMAIL THIS PAGE TO A FRIEND
36946 Sigma-Aldrich

Naphthalene solution

100 ng/μL in cyclohexane, analytical standard

Purchase

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1145 - class 3 - PG 2 - Cyclohexane, solution
WGK Germany 
2
Flash Point(F) 
-.4 °F
Flash Point(C) 
-18 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?