• USA Home
  • 36953 - Benzo[ghi]perylene solution

EMAIL THIS PAGE TO A FRIEND
36953 Sigma-Aldrich

Benzo[ghi]perylene solution

100 ng/μL in cyclohexane, analytical standard

  • CAS Number 191-24-2

  • Empirical Formula (Hill Notation) C22H12

  • Molecular Weight 276.33

  •  Beilstein Registry Number 1913029

  •  MDL number MFCD00004135

  •  PubChem Substance ID 329755552

Purchase

Properties

Related Categories A-B, Additional Standards, All Polycyclic Aromatic Hydrocarbon (PAH) Standards, Alpha Sort, Analytical Standards,
grade   analytical standard
InChI Key   GYFAGKUZYNFMBN-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
concentration   100 ng/μL in cyclohexane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1145 3 / PGII
WGK Germany 
2
Flash Point(F) 
-.4 °F
Flash Point(C) 
-18 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?