• USA Home
  • 36959 - Acenaphthene solution

EMAIL THIS PAGE TO A FRIEND
36959 Sigma-Aldrich

Acenaphthene solution

100 μg/mL in cyclohexane, analytical standard

  • CAS Number 83-32-9

  • Empirical Formula (Hill Notation) C12H10

  • Molecular Weight 154.21

  •  Beilstein Registry Number 386081

  •  MDL number MFCD00003807

  •  PubChem Substance ID 329755555

Purchase

Properties

Related Categories A-B, Additional Standards, All Polycyclic Aromatic Hydrocarbon (PAH) Standards, Alpha Sort, Analytical Standards,
grade   analytical standard
InChI Key   CWRYPZZKDGJXCA-UHFFFAOYSA-N
shelf life   limited shelf life, expiry date on the label
concentration   100 μg/mL in cyclohexane
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN1145 - class 3 - PG 2 - Cyclohexane, solution
WGK Germany 
2
Flash Point(F) 
-.4 °F
Flash Point(C) 
-18 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?