EMAIL THIS PAGE TO A FRIEND
36986 Sigma-Aldrich

Iprobenfos solution

100 ng/μL in methanol, PESTANAL®, analytical standard

  • CAS Number 26087-47-8

  • Empirical Formula (Hill Notation) C13H21O3PS

  • Molecular Weight 288.34

  •  Beilstein Registry Number 1974687

  •  EC Number 200-659-6

  •  MDL number MFCD00191226

  •  PubChem Substance ID 329755564

Purchase

Properties

Related Categories All Fungicides, Alpha Index of All Pesticide Standards, Analytical Standards, Analytical/Chromatography, Fungicides,
grade   analytical standard
InChI Key   FCOAHACKGGIURQ-UHFFFAOYSA-N
product line   PESTANAL®
shelf life   limited shelf life, expiry date on the label
concentration   100 ng/μL in methanol
application(s)   HPLC: suitable
  gas chromatography (GC): suitable
format   single component solution
storage temp.   2-8°C

Description

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 1230 3/PG 2
WGK Germany 
1
Flash Point(F) 
51.8 °F
Flash Point(C) 
11 °C

Documents

Certificate of Analysis

Protocols & Articles

Articles

GC Analysis of 'Positive List' Chemicals (Mix 4) on SLB®-5ms

From our library of Articles, Sigma-Aldrich presents GC Analysis of 'Positive List' Chemicals (Mix 4) on SLB®-5ms
Keywords: Chromatography, Gas chromatography, Herbicides, Pesticides, Purification

LC/MS/MS of Pesticides in Oranges on Ascentis® Express C18 after Sample Prep using Supel™ Que Z-Sep/C18 QuEChERS sorbent

From our library of Articles, Sigma-Aldrich presents LC/MS/MS of Pesticides in Oranges on Ascentis® Express C18 after Sample Prep using Supel™ Que Z-Sep/C18 QuEChERS sorbent
Keywords: Chromatography, High performance liquid chromatography, Liquid chromatography mass spectrometry, Mass spectrometry, Pesticides, Sample preparations, Solid phase extractions

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?