• USA Home
  • 37028 - Special indicator paper

EMAIL THIS PAGE TO A FRIEND
37028 Sigma-Aldrich

Special indicator paper

non-bleeding, pH 4.5-10.0

Synonym: Indicator sticks, special

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Test Papers/Sticks, pH Papers/Sticks More...
quality   non-bleeding
pH   4.5-10.0

Description

Other Notes

Same product — same quality — now Sigma-Aldrich brand

Packaging

box with 100 strips

General description

Special indicator papers cover range from 2 to 5 pH units in general. It has the capacity to change colour at particular pH value. Nonbleeding indicator papers are prepared by either direct dyes or reactive dyes. During reactive dyes procedure, pure linters are suspended in water and reactive dye is added. The mixture is made alkaline for the reaction to start. The fiber pulp is centrifuged, washed to be electrolyte-free and processed as pH paper. Paper produced in this way is bonded to plastic material in order to be used as stick or strip.

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Set your institution to view full text papers.
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?