• USA Home
  • 37065 - Special indicator paper

EMAIL THIS PAGE TO A FRIEND
37065 Sigma-Aldrich

Special indicator paper

pH 6.4-8.0

Synonym: Indicator paper, special

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Application Specific Reagents, Test Papers/Sticks, pH Papers/Sticks More...
pH   6.4-8.0
suitability   corresponds for shade of color acc. to scale

Description

Packaging

6 boxes with 1 color chart
roll with approx. 5 m

Price and Availability

Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

37110 pH-Indicator strips, pH-range 6.0 - 8.1

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?