EMAIL THIS PAGE TO A FRIEND
37110 Sigma-Aldrich
pH-Indicator strips
pH-range 6.0 - 8.1
Purchase
Description
Features and Benefits
Each strip has an identification scale and printed pH scale. The strips can be utilized in colored solutions and suspensions with very high precision.
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
Packages of 200 measuring strips.
Application
pH indicator strip may be used during the synthesis of modified nucleotides, for site-directed DNA photoaffinity labelling of RNA polymerase III transcription complexes.
General description
Indicator papers are prepared by either direct dyes or reactive dyes. During reactive dyes procedure, pure linters are suspended in water and reactive dye is added. The mixture is made alkaline for the reaction to start. The fiber pulp is centrifuges, washed to be electrolyte-free and processed as pH paper. Paper produced in this way is bonded to plastic material in order to be used as stick or strip. Some strips are produced by impregnation which bleed, certain portion of the indicator dye dissolves in the test solution during immersion. pH strips with covalently bonded indicator dyes remain indefinitely in solution without migration or dissolution.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?