EMAIL THIS PAGE TO A FRIEND
37200 Sigma-Aldrich

Dihexylamine

purum, ≥97.0% (GC)

  • CAS Number 143-16-8

  • Linear Formula [CH3(CH2)5]2NH

  • Molecular Weight 185.35

  •  Beilstein Registry Number 1738519

  •  EC Number 205-588-4

  •  MDL number MFCD00009521

Purchase

Properties

vapor density   6.4 (vs air)
InChI Key   PXSXRABJBXYMFT-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
refractive index   n20/D 1.432(lit.)
  n20/D 1.433
bp   192-195 °C(lit.)
density   0.795 g/mL at 25 °C(lit.)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN2922 - class 8 - PG 2 - EHS - acidic - Corrosive liquids, toxic, n.o.s., HI: all
WGK Germany 
2
RTECS 
IH6600000
Flash Point(F) 
203 °F
Flash Point(C) 
95 °C
Protocols & Articles
Related Products

replaced by

Product #

Description

Add to Cart

131202 Dihexylamine, 97%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?