• USA Home
  • 37203 - Ascorbic acid test sticks Quantofix®

EMAIL THIS PAGE TO A FRIEND
37203 Sigma-Aldrich

Ascorbic acid test sticks Quantofix®

50-2000 mg/L, (vitamin C)

Purchase

Description

General description

Contents:
Tube with 100 test sticks.

Storage:
Avoid exposing the strips to sunlight and moisture. Keep container cool and dry (store below +30 °C).

Gradation:
0 - 50 - 100 - 200 - 300 - 500 - 1000 - 2000 mg/L Ascorbic acid / Vitamin C

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
nwg

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?