• USA Home
  • 37207 - Aluminum test kit Quantofix®

EMAIL THIS PAGE TO A FRIEND
37207 Sigma-Aldrich

Aluminum test kit Quantofix®

5-500 mg/L, (Al3+)

Purchase

Description

General description

Contents:
Kit including reagents and test sticks for 100 tests.

Storage:
Avoid exposing the sticks to sunlight and moisture. Store the kit below +30 °C in a dry place.

Gradation:
0 - 5 - 20 - 50 - 200 - 500 mg/L Al3+

Legal Information

Quantofix is a registered trademark of Macherey-Nagel GmbH & Co., KG

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?