• USA Home
  • 37360 - 3,4-Dihydro-2-pyridol[1,2-a]pyrimidinone

EMAIL THIS PAGE TO A FRIEND
37360 Sigma-Aldrich

3,4-Dihydro-2-pyridol[1,2-a]pyrimidinone

for GC derivatization, ≥99.0%

  • CAS Number 5439-14-5

  • Empirical Formula (Hill Notation) C8H8N2O

  • Molecular Weight 148.16

  •  Beilstein Registry Number 3667723

  •  EC Number 226-618-2

  •  MDL number MFCD00042798

  •  PubChem Substance ID 329755640

Purchase

Properties

Related Categories Alkylation Reagents, Analytical Reagents, Analytical/Chromatography, Building Blocks, C6 to C8,
grade   for GC derivatization
InChI Key   XQIOBBHIEUGFCI-UHFFFAOYSA-N
assay   ≥99.0% (NT)
  ≥99.0%
mp   185-187 °C
storage temp.   2-8°C

Description

Other Notes

A very effective non-basic hydrogen halide captor in various reactions; Proton sponge for glycosylations

Packaging

10 g in glass bottle

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

08168 Timestrip Plus 8 °C

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?