• USA Home
  • 37408 - AQUANAL-plus nitrate (NO3) 5-140 mg/L
EMAIL THIS PAGE TO A FRIEND
37408 Sigma-Aldrich
AQUANAL-plus nitrate (NO3) 5-140 mg/L
test set with color comparator and check solution
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items, Tests for semi-quantitative determination (color charts) More...
usage   sufficient for 200 tests
measuring range   10-150 mg/L, (photometric, Spectro 3)
  5-140 mg/L, (color chart)
Description
Analysis Note
Method
Formation of a yellowbrown azo dye with gentisic acid
Gradation (color chart)
0 - 5 - 10 - 20 - 40 - 60 - 80 - 100 - 120 - 140 mg/L (NO3-)
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Signal word 
Danger
RIDADR 
UN 3316 9
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replacement part
Product #
Description
Add to Cart
37448 AQUANAL-plus nitrate (NO3) 5-140 mg/L, refill pack for 37408
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?