• USA Home
  • 37412 - AQUANAL-plus silicon (Si) 0.01-0.25 mg/L
EMAIL THIS PAGE TO A FRIEND
37412 Sigma-Aldrich
AQUANAL-plus silicon (Si) 0.01-0.25 mg/L
test set with color comparator and check solution
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items, Tests for semi-quantitative determination (color charts) More...
usage   sufficient for 150 tests
measuring range   0.005-2 mg/L, (photometric, Spectro 3)
  0.01-0.25 mg/L, (color chart)
Description
Analysis Note
Method
Water-soluble silicon (silicic acid) forms with ammonium molybdate a blue color complex (silicomolybdenum blue)
Gradation (color chart)
0 - 0.01 - 0.02 - 0.04 - 0.06 - 0.08 - 0.10 - 0.15 - 0.20 - 0.25 mg/L (Si)
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replacement part
Product #
Description
Add to Cart
37452 AQUANAL-plus silicon (Si) 0.01-0.25 mg/L, refill pack for 37412
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?