• USA Home
  • 37413 - AQUANAL-plus zinc (Zn) 0.1-5.0 mg/L
EMAIL THIS PAGE TO A FRIEND
37413 Sigma-Aldrich
AQUANAL-plus zinc (Zn) 0.1-5.0 mg/L
test set with color comparator and check solution
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items, Tests for semi-quantitative determination (color charts) More...
usage   sufficient for 120 tests
measuring range   0.1-5.0 mg/L, (color chart)
Description
Analysis Note
Method
Formation of a green color complex with brilliant green
Gradation (color chart)
0 - 0.1 - 0.2 - 0.3 - 0.4 - 0.5 - 0.7 - 1.0 - 2.0 - 5.0 mg/L (Zn2+)
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Signal word 
Danger
Hazard statements 
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN 3316 9
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replacement part
Product #
Description
Add to Cart
37453 AQUANAL-plus zinc (Zn) 0.1-5.0 mg/L, refill pack for 37413
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?