• USA Home
  • 37418 - AQUANAL-plus oxygen scavenger
EMAIL THIS PAGE TO A FRIEND
37418 Sigma-Aldrich
AQUANAL-plus oxygen scavenger
test set with color comparator
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items, Tests for semi-quantitative determination (color charts) More...
usage   sufficient for 100 tests
measuring range   0.2-2 mg/L, (photometric, Spectro 3)
  0.2-3.0 mg/L
Description
Analysis Note
Method
Formation of a blue color complex with 2,4,6-tris(2-pyridyl)-1,3,5-triazine
Gradation (color chart)
0 - 0.2 - 0.5 - 0.8 - 1.2 - 1.6 - 2.0 - 3.0 mg/L
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
WGK Germany 
2
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replacement part
Product #
Description
Add to Cart
37458 AQUANAL-plus oxygen scavenger, refill pack for 37418
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?