• USA Home
  • 37421 - AQUANAL-plus iron (Fe) 0.02-0.2 mg/L
EMAIL THIS PAGE TO A FRIEND
37421 Sigma-Aldrich
AQUANAL-plus iron (Fe) 0.02-0.2 mg/L
test set with color comparator and check solution
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
usage   sufficient for 100 tests
measuring range   0.02-0.2 mg/L, (color chart)
  0.02-0.3 mg/L, (photometric, Spectro 3)
Description
Analysis Note
Method
Formation of a blue color complex with 2,4,6-tri-2-pyridyl-1,3,5-triazine
Gradation (color chart)
0 - 0.02 - 0.04 - 0.06 - 0.08 - 0.10 - 0.13 - 0.16 - 0.20 mg/L (Fe2+/3+)
Other Notes
Same product — same quality — now Sigma-Aldrich brand
General description
Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
replacement part
Product #
Description
Add to Cart
37461 AQUANAL-plus iron (Fe) 0.02-0.2 mg/L, refill pack for 37421
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?