• USA Home
  • 37423 - AQUANAL-plus phosphate (PO4) 3-120 mg/L
EMAIL THIS PAGE TO A FRIEND
37423 Sigma-Aldrich
AQUANAL-plus phosphate (PO4) 3-120 mg/L
test set with color comparator and check solution
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items, Tests for semi-quantitative determination (color charts) More...
usage   sufficient for 210 tests
measuring range   1-150 mg/L, (photometric, Spectro 3)
  3-120 mg/L, (color chart)
Description
Analysis Note
Method
Formation of a orange-yellow color complex with ammonium molybdate and ammonium vanadate
Principle
DEV
Gradation (color chart)
0 - 3 - 6 - 10 - 18 - 30 - 60 - 120 mg/L (PO43-)
Other Notes
Same product — same quality — now Sigma-Aldrich brand
General description
Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
WGK Germany 
2
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?