• USA Home
  • 37428 - AQUANAL-plus oxygen (O2) 1-12 mg/L

EMAIL THIS PAGE TO A FRIEND
37428 Sigma-Aldrich

AQUANAL-plus oxygen (O2) 1-12 mg/L

test set with color comparator

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items, Tests for semi-quantitative determination (color charts) More...
InChI Key   MYMOFIZGZYHOMD-UHFFFAOYSA-N
usage   sufficient for 30 tests
measuring range   0.5-12 mg/L, (photometric, Spectro 3)
  1-12 mg/L, (color chart)

Description

Analysis Note

Method
modified method according to WInkler

Gradation (color chart)
0 - 1 - 3 - 5 - 7 - 9 - 12 mg/L (O2)

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replacement part

Product #

Description

Add to Cart

32021 AQUANAL-plus oxygen (O2) 1-12 mg/L, refill pack for 37428

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?