• USA Home
  • 37429 - AQUANAL-plus sulfate (SO4) 50-330 mg/L

EMAIL THIS PAGE TO A FRIEND
37429 Sigma-Aldrich

AQUANAL-plus sulfate (SO4) 50-330 mg/L

test set with color comparator and check solution

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items, Tests for semi-quantitative determination (color charts) More...
InChI Key   QAOWNCQODCNURD-UHFFFAOYSA-L
usage   sufficient for 100 tests
measuring range   50-330 mg/L, (color chart)
  50-400 mg/L, (photometric, Spectro 3)

Description

Analysis Note

Method
Forming of a red-violet colored chloroanilate complex

Gradation (color chart)
0 - 50 - 80 - 110 - 140 - 180 - 240 - 330 mg/L (SO42-)

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
RIDADR 
UN 3316 9

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?