• USA Home
  • 37432 - AQUANAL-plus cadmium (Cd) 0.02-1.2 mg/L

EMAIL THIS PAGE TO A FRIEND
37432 Sigma-Aldrich

AQUANAL-plus cadmium (Cd) 0.02-1.2 mg/L

test set with color comparator

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items, Tests for semi-quantitative determination (color charts) More...
InChI Key   BDOSMKKIYDKNTQ-UHFFFAOYSA-N
usage   sufficient for 120 tests
measuring range   0.01-2 mg/L, (photometric, Spectro 3)
  0.02-1.2 mg/L, (color chart)

Description

Analysis Note

Method
Formation of a yellow-red color complex with cadion

Gradation (color chart)
0 - 0.02 - 0.05 - 0.1 - 0.15 - 0.2 - 0.3 - 0.5 - 0.8 - 1.2 mg/L (Cd2+)

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replacement part

Product #

Description

Add to Cart

37472 AQUANAL-plus cadmium (Cd) 0.02-1.2 mg/L, refill pack for 37432

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?