• USA Home
  • 37433 - AQUANAL-plus hydrazine (N2H4) 0.02-1.0 mg/L
EMAIL THIS PAGE TO A FRIEND
37433 Sigma-Aldrich
AQUANAL-plus hydrazine (N2H4) 0.02-1.0 mg/L
test set with color comparator
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items, Tests for semi-quantitative determination (color charts) More...
usage   sufficient for 130 tests
measuring range   0.01-2 mg/L, (photometric, Spectro 3)
  0.02-1.0 mg/L, (color chart)
Description
Analysis Note
Method
Formation of a yellow color complex with 4-(dimethylamino)benzaldehyde
Principle
DEV
Gradation (color chart)
0 - 0.02 - 0.05 - 0.08 - 0.12 - 0.25 - 0.5 - 0.75 - 1.0 mg/L (N2H2)
Other Notes
Same product — same quality — now Sigma-Aldrich brand
General description
Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations.
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
replacement part
Product #
Description
Add to Cart
37473 AQUANAL-plus hydrazine (N2H4) 0.02-1.0 mg/L, refill pack for 37433
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?