• USA Home
  • 37447 - AQUANAL-plus nickel (Ni) 0.02-0.5 mg/L

EMAIL THIS PAGE TO A FRIEND
37447 Sigma-Aldrich

AQUANAL-plus nickel (Ni) 0.02-0.5 mg/L

refill pack for 37407

Purchase

Description

Analysis Note

also for photometric evaluation

General description

Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN 3316 9
Flash Point(F) 
68 °F
Flash Point(C) 
20 °C

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?