EMAIL THIS PAGE TO A FRIEND
37478 Sigma-Aldrich

5-Methoxyresorcinol

purum, ≥97.0% (GC)

Synonym: 3,5-Dihydroxyanisole, Phloroglucinol monomethyl ether

  • CAS Number 2174-64-3

  • Linear Formula CH3OC6H3-1,3-(OH)2

  • Molecular Weight 140.14

  •  Beilstein Registry Number 1423578

  •  EC Number 218-532-9

  •  MDL number MFCD00002285

  •  PubChem Substance ID 329755653

Purchase

Properties

Related Categories Building Blocks, Chemical Synthesis, Organic Building Blocks, Oxygen Compounds, Polyols More...
InChI Key   HDVRLUFGYQYLFJ-UHFFFAOYSA-N
grade   purum
assay   ≥97.0% (GC)
bp   188-189 °C/12 mmHg(lit.)
mp   76-78 °C
  78-80 °C(lit.)
  83-85 °C (2nd modification)

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
Personal Protective Equipment 
RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replaced by

Product #

Description

Add to Cart

232491 5-Methoxyresorcinol, 98%

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?