• USA Home
  • 37488 - AQUANAL-plus nitrate (NO3) 5-140 mg/L

EMAIL THIS PAGE TO A FRIEND
37488 Sigma-Aldrich

AQUANAL-plus nitrate (NO3) 5-140 mg/L

check solution for 37408 (calibrated standard (NO3 = 2 mg/mL))

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
grade   check solution for 37408 (calibrated standard (NO3 = 2 mg/mL))

Description

Application

AQUANAL-plus nitrate (NO3) may be used as a calibrated standard during an experimental study conducted to study the mechanism of graphite oxide (GO) thermal reduction.

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?