• USA Home
  • 37493 - AQUANAL-plus zinc (Zn) 0.1-5.0 mg/L

EMAIL THIS PAGE TO A FRIEND
37493 Sigma-Aldrich

AQUANAL-plus zinc (Zn) 0.1-5.0 mg/L

check solution for 37413 (calibrated standard (Zn = 0.025 mg/mL))

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
grade   check solution for 37413 (calibrated standard (Zn = 0.025 mg/mL))

Description

Packaging

30 mL

General description

Aquanal kits can be used for the determination of hardness and phosphate ammonia, nitrate, and nitrite concentrations.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Danger
Hazard statements 
RIDADR 
UN3264 - class 8 - PG 2 - EHS - Corrosive liquid, acidic, inorganic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?