• USA Home
  • 37537 - AQUANAL-plus gold (Au) 0.1-3.0 mg/L

EMAIL THIS PAGE TO A FRIEND
37537 Sigma-Aldrich

AQUANAL-plus gold (Au) 0.1-3.0 mg/L

reagents for photometric analysis

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
grade   reagents for photometric analysis
usage   60 tests
measuring range   0.1-5 mg/L, (photometric, Spectro 3)

Description

Analysis Note

forming of a red colored complex with promazine hydrochloride in a acidly medium

General description

Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS05  GHS05
Signal word 
Danger
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?