• USA Home
  • 37547 - AQUANAL pooltester chlorine/pH
EMAIL THIS PAGE TO A FRIEND
37547 Sigma-Aldrich
AQUANAL pooltester chlorine/pH
refill pack for 37546
Purchase
Description
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
30 tests chlorine and pH-value each
General description
Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations. AQUANAL-Pooltester is designed for the measurement of pH and chlorine levels in swimming pools.
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
NONH for all modes of transport
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?