• USA Home
  • 37557 - AQUANAL-Oekotest Water Laboratory

EMAIL THIS PAGE TO A FRIEND
37557 Sigma-Aldrich

AQUANAL-Oekotest Water Laboratory

sufficient reagents for 50 to 60 analyses (of ammonium, nitrate, nitrite, phosphate, total hardness and pH-value)

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Kits/Cases, Potable- & Waste-Water Testing
grade   sufficient reagents for 50 to 60 analyses (of ammonium, nitrate, nitrite, phosphate, total hardness and pH-value)

Description

General description

Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
UN 3316 9
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

replacement part

Product #

Description

Add to Cart

37558 AQUANAL-Oekotest Water Laboratory, refill pack for 37557
37574 AQUANAL reagent for the determination of total hardness, one-component reagent, 1 drop = 1 German degree of hardness
37541 AQUANAL-Oekotest Water Laboratory, cuvettes refill for 37557
37555 AQUANAL-Oekotest water laboratory pH-value reagent
37606 AQUANAL Oekotest Nitrite, refill for 37557 + 37566
37599 AQUANAL Oekotest Nitrate, refill for 37557 + 37566
37607 AQUANAL Oekotest Phosphate, refill for 37557 + 37556

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?