• USA Home
  • 37562 - AQUANAL-plus Compact Lab

EMAIL THIS PAGE TO A FRIEND
37562 Sigma-Aldrich

AQUANAL-plus Compact Lab

Purchase

Description

Application

for photometric determination of 17 environmental parameters in drinking water and in weakly contamined waste water
determination only with Spectro Photometer possible
The photometer must be ordered separately

General description

Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations. AQUANAL-Compact Labs are collections of test kits, reagents, photometers and other accessories that provide all you need to perform detailed analysis of specific types of water samples. It allows rapid, reliable water analysis. The product is ideally suited to meet the requirements of emergency responders, like fire brigades, police, environ-mental protection officers and local authorities. It is also useful for educational purposes. It was made for the measurement of seventeen pollutants in drinking water.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Kit component also available separately

Description

Product #

Add to Cart

AQUANAL-plus ammonium (NH4) 0.2-8.0 mg/L, refill pack for 37400 SDS 37440
AQUANAL-plus chlorine (Cl2) free and bound 0.01-0.30 mg/L, refill pack for 37419 SDS 37459
AQUANAL-plus chloride (Cl) 5-300 mg/L, refill pack for 37401 SDS 37441
AQUANAL-plus chromium (Cr) 0.005-0.1 mg/L, refill pack for 37402 SDS 37442
AQUANAL-plus cyanide (CN) 0.03-0.7 mg/L, refill pack for 37403 SDS 37443
AQUANAL-plus iron (Fe) 0.2-15 mg/L, refill pack for 37404 SDS 37444
AQUANAL reagent for the determination of total hardness, one-component reagent, 1 drop = 1 German degree of hardness SDS 37574
AQUANAL-plus copper (Cu) 0.05-0.5 / 0.4-4.5 mg/L, refill pack for 37424 SDS 37464
AQUANAL-plus nickel (Ni) 0.02-0.5 mg/L, refill pack for 37407 SDS 37447
AQUANAL-plus nitrate (NO3) 1-50 mg/L, refill pack for 37409 SDS 37449
AQUANAL-plus nitrite (NO2) 0.005-0.1 mg/L, refill pack for 37410 SDS    
AQUANAL-plus phenols 0.1-3.0 mg/L, refill pack for 37427 SDS 37467
AQUANAL-plus phosphate (PO4) 0.02-0.4 mg/L, refill pack for 37411 SDS 37451
Special indicator paper, non-bleeding, pH 4.5-10.0 SDS 37028
AQUANAL-plus oxygen (O2) 1-12 mg/L, refill pack for 37428 SDS    
AQUANAL-plus sulfate (SO4) 50-330 mg/L, refill pack for 37429 SDS 37469
AQUANAL-plus sulfide (S2-) 0.03-0.8 mg/L, refill pack for 37430 SDS    
Safety & Documentation

Safety Information

Signal word 
Danger
Precautionary statements 
Supplemental Hazard Statements 
Contact with acids liberates toxic gas.
RIDADR 
UN 3316 9

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?