• USA Home
  • 37563 - AQUANAL analytical case for building restoration
EMAIL THIS PAGE TO A FRIEND
37563 Sigma-Aldrich
AQUANAL analytical case for building restoration
total set of reagents (for analysis of chloride, total hardness, magnesium, nitrate, sulfate and sulfide in scumming of brickwork)
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Kits/Cases, Potable- & Waste-Water Testing
grade   total set of reagents (for analysis of chloride, total hardness, magnesium, nitrate, sulfate and sulfide in scumming of brickwork)
Description
Analysis Note
determination only with Spectro Photometer possible
The photometer must be ordered separately
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Signal word 
Danger
Supplemental Hazard Statements 
Contact with acids liberates very toxic gas.
RIDADR 
UN 3316 9
WGK Germany 
3
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?