• USA Home
  • 37566 - AQUANAL Fishwater Lab
EMAIL THIS PAGE TO A FRIEND
37566 Sigma-Aldrich
AQUANAL Fishwater Lab
total set of reagents (for the fast analysis of ammonium, nitrate, nitrite, phosphate, oxygen, iron, carbonate hardness, pH and total hardness in water)
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Kits/Cases, Potable- & Waste-Water Testing
grade   total set of reagents (for the fast analysis of ammonium, nitrate, nitrite, phosphate, oxygen, iron, carbonate hardness, pH and total hardness in water)
Description
Other Notes
Same product — same quality — now Sigma-Aldrich brand
General description
Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations. Nine parameters which are significant for the quality of an aqueous environment (e.g. aquaculture, lakes, rivers, ponds and aquariums) can be determined directly using AQUANAL Fishwater Lab. Fishwater Lab is designed for the measurement of the most important water quality parameters in aquariums and fish farms.
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
Signal word 
Danger
Precautionary statements 
RIDADR 
UN 3316 9
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
replacement part
Product #
Description
Add to Cart
37028 Special indicator paper, non-bleeding, pH 4.5-10.0
37574 AQUANAL reagent for the determination of total hardness, one-component reagent, 1 drop = 1 German degree of hardness
37542 AQUANAL Fishwater Lab, cuvettes refill pack for 37566
37596 AQUANAL Fish carbonate hardness, refill for 37566
37595 AQUANAL Oekotest Ammonium, refill for 37557 + 37566
37606 AQUANAL Oekotest Nitrite, refill for 37557 + 37566
37599 AQUANAL Oekotest Nitrate, refill for 37557 + 37566
37607 AQUANAL Oekotest Phosphate, refill for 37557 + 37556
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?