• USA Home
  • 37574 - AQUANAL reagent for the determination of total hardness

EMAIL THIS PAGE TO A FRIEND
37574 Sigma-Aldrich

AQUANAL reagent for the determination of total hardness

one-component reagent, 1 drop = 1 German degree of hardness

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
grade   one-component reagent

Description

General description

Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations. Total Hardness is essentially determined by the calcium salts dissolved in water. The presence of a high calcium content in water corresponds to a high water hardness measured in German degrees of hardness (°dH). The performance characteristic of water is, however, significantly influenced by the hardness. Basically, soft water (e.g. calcium-deficient water) is more suitable for hot water applications: coffee machines, washing machines, steam irons, kettles and water heaters can be contaminated by hard water and possibly damaged. Many manufacturers give recommendations for regular cleanings or application of water additives for hard water.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Symbol 
GHS07  GHS07
Signal word 
Warning
Hazard statements 
Precautionary statements 
RIDADR 
NONH for all modes of transport
WGK Germany 
2

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?