• USA Home
  • 37736 - AQUANAL-professional tube test COD

EMAIL THIS PAGE TO A FRIEND
37736 Sigma-Aldrich

AQUANAL-professional tube test COD

0-150 mg/L

Synonym: COD cuvette tests

Purchase

Properties

Related Categories Analytical Reagents, Analytical/Chromatography, Kits/Cases, Potable- & Waste-Water Testing
measuring range   0-150 mg/L

Description

Analysis Note

oxidation by potassium dichromate in sulfuric acid solution

General description

Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations. AQUANAL-professional tube tests were developed to provide a fast safe and clean method in order to perform water analysis in sewage treatment plants. The sample is pipetted into a tube that contains the test reagent. For some tests, an additional reagent is required that has to be added manually. The COD test is shipped in ready-to-use solutions.

Legal Information

AQUANAL is a trademark of Sigma-Aldrich International GmbH

Price and Availability

Safety & Documentation

Safety Information

Signal word 
Danger
Hazard statements 
RIDADR 
UN2922 - class 8 - PG 2 - EHS - acidic - Corrosive liquids, toxic, n.o.s., HI: all
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?