• USA Home
  • 37739 - AQUANAL-professional Ammonia Tube test
EMAIL THIS PAGE TO A FRIEND
37739 Sigma-Aldrich
AQUANAL-professional Ammonia Tube test
Purchase
Properties
Related Categories Analytical Reagents, Analytical/Chromatography, Potable- & Waste-Water Testing, Replacement Kit Items
measuring range   0.01-50 mg/L, (photometric, Spectro 3, Spectro 1000)
Description
Analysis Note
Method: Indophenol blue reacation
Other Notes
Same product — same quality — now Sigma-Aldrich brand
Packaging
24 reaction tubes, 1 blank tube
General description
Aquanal kits can be used for the determination of hardness and phosphate, ammonia, nitrate, and nitrite concentrations. AQUANAL-professional tube tests were developed to provide a fast safe and clean method in order to perform water analysis in sewage treatment plants. The sample is pipetted into a tube that contains the test reagent. For some tests, an additional reagent is required that has to be added manually.
Legal Information
AQUANAL is a trademark of Sigma-Aldrich International GmbH
Price and Availability
Safety & Documentation
Safety Information
RIDADR 
UN 3316 9
Documents
Certificate of Analysis
Protocols & Articles
Peer-Reviewed Papers
15
References
Set your institution to view full text papers.
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?